Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-5-iodobenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

69178-1G 69178-25G 69178-500MG 69178-5G
76905-750EA 258.86 USD InStock
76905-750 76905-752 76905-754 76905-756
2,4-Difluoro-5-iodobenzaldehyde ≥97% research grade
2,4-Difluoro-5-iodobenzaldehyde


Formula: C₇H₃F₂IO
MW: 267.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789181
CAS Number: 1097626-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty