Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-cyano-5-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

80985-1G 80985-500MG 80985-5G
76905-970EA 970.73 USD InStock
76905-970 76905-972 76905-974
2-Chloro-4-cyano-5-methylbenzoic acid ≥97%
2-Chloro-4-cyano-5-methylbenzoic acid


Formula: C₉H₆ClNO₂
Storage Temperature: Ambient
CAS Number: 1805497-46-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty