Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diisopropylbenzoic Acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

90597-1G 90597-25G 90597-500MG 90597-5G
76906-062EA 138.98 USD InStock
76906-062 76906-064 76906-066 76906-068
2,6-Diisopropylbenzoic Acid ≥97% research grade
2,6-Diisopropylbenzoic Acid


Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09965980
CAS Number: 92035-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty