Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Bis(benzyloxy)-5-bromopyrimidine ≥97% research grade

Supplier: AOB CHEM USA
Warning

10433-25G 10433-5G
76924-320EA 194.15 USD InStock
76924-320 76924-322
2,4-Bis(benzyloxy)-5-bromopyrimidine ≥97% research grade
2,4-Bis(benzyloxy)-5-bromopyrimidine


Formula: C₁₈H₁₅BrN₂O₂
MW: 371.2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00267932
CAS Number: 41244-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty