Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formyl-3-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

10474-1G 10474-25G 10474-500MG 10474-5G
76924-342EA 231.28 USD InStock
76924-342 76924-344 76924-346 76924-348
4-Formyl-3-methylphenylboronic acid ≥97%
4-Formyl-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD10566601
CAS Number: 398151-59-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty