Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-isopropoxybenzeneboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

12066-1G 12066-25G 12066-5G
76924-662EA 62.6 USD InStock
76924-662 76924-664 76924-666
4-Fluoro-2-isopropoxybenzeneboronic acid ≥97% research grade
4-Fluoro-2-isopropoxybenzeneboronic acid


Formula: C₉H₁₂BFO₃
MW: 198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411287
CAS Number: 480438-59-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty