Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isobutylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

12134-25G 12134-5G
76924-692EA 375.56 USD InStock
76924-692 76924-694
4-Isobutylphenylboronic acid pinacol ester ≥97%
4-Isobutylphenylboronic acid pinacol ester


Formula: C₁₆H₂₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD05663842
CAS Number: 1033753-01-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty