Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

14014-1G 14014-25G 14014-500MG 14014-5G
76924-938EA 147.47 USD InStock
76924-938 76924-940 76924-942 76924-944
3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid ≥97%
3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235046
CAS Number: 874219-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty