Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-ethylbenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

14044-1G 14044-25G 14044-500MG 14044-5G
76925-004EA 144.29 USD InStock
76925-004 76925-006 76925-008 76925-010
4-Bromo-2-ethylbenzaldehyde ≥97%
4-Bromo-2-ethylbenzaldehyde


Formula: C₉H₉BrO
Storage Temperature: Refrigerator
MDL Number: MFCD09835094
CAS Number: 1114808-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty