Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formyl-3-methylthiophene-2-boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

15530-1G 15530-25G 15530-500MG 15530-5G
76925-464EA 195.21 USD InStock
76925-464 76925-466 76925-468 76925-470
5-Formyl-3-methylthiophene-2-boronic acid ≥97%
5-Formyl-3-methylthiophene-2-boronic acid
Precaución: for Research Use Only


Formula: C₆H₇BO₃S
MW: 169.994 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06657888
CAS Number: 1072952-28-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty