Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-2-bromobenzotrifluoride ≥95%

Supplier: AOB CHEM USA
Warning

15537-1G 15537-25G 15537-500MG 15537-5G
76925-474EA 168.69 USD InStock
76925-474 76925-476 76925-478 76925-480
5-(Benzyloxy)-2-bromobenzotrifluoride ≥95%
5-(Benzyloxy)-2-bromobenzotrifluoride
Precaución: for Research Use Only


Formula: C₁₄H₁₀BrF₃O
MW: 331.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195732
CAS Number: 678164-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty