Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-formyl-5-methylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

15888-1G 15888-25G 15888-5G
76925-572EA 96.55 USD InStock
76925-572 76925-574 76925-576
4-Fluoro-3-formyl-5-methylphenylboronic acid ≥97% research grade
4-Fluoro-3-formyl-5-methylphenylboronic acid


Formula: C₈H₈BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11111909
CAS Number: 1310384-23-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty