Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Acetyl-2-fluorobenzeneboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

16467-25G 16467-5G
76925-768EA 297.06 USD InStock
76925-768 76925-770
5-Acetyl-2-fluorobenzeneboronic acid pinacol ester ≥97% research grade
5-Acetyl-2-fluorobenzeneboronic acid pinacol ester


Formula: C₁₄H₁₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD16996292
CAS Number: 765916-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty