Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-6-methoxybenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

16493-1G 16493-25G 16493-5G
76925-784EA 114.58 USD InStock
76925-784 76925-786 76925-788
4-Bromo-2-fluoro-6-methoxybenzaldehyde ≥97% research grade
4-Bromo-2-fluoro-6-methoxybenzaldehyde


Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543644
CAS Number: 856767-09-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty