Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tavaborole ≥97%

Supplier: AOB CHEM USA
Warning

16685-1G 16685-25G 16685-5G
76925-874EA 79.57 USD InStock
76925-874 76925-876 76925-878
Tavaborole ≥97%
Tavaborole
Precaución: for Research Use Only


Formula: C₇H₆BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD10699483
CAS Number: 174671-46-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty