Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formyl-2,6-dimethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16696-1G 16696-25G 16696-500MG 16696-5G
76925-886EA 135.8 USD InStock
76925-886 76925-888 76925-890 76925-892
4-Formyl-2,6-dimethylphenylboronic acid ≥97%
4-Formyl-2,6-dimethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD19105353
CAS Number: 1228829-13-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty