Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Acetyl-3-thienylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

17234-1G 17234-25G 17234-500MG 17234-5G
76926-096EA 231.28 USD InStock
76926-096 76926-098 76926-100 76926-102
5-Acetyl-3-thienylboronic acid ≥97%
5-Acetyl-3-thienylboronic acid
Precaución: for Research Use Only


Formula: C₆H₇BO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD08701784
CAS Number: 942190-74-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty