Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methoxyisonicotinaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

17649-1G 17649-25G 17649-500MG 17649-5G
76926-256EA 159.14 USD InStock
76926-256 76926-258 76926-260 76926-262
5-Chloro-2-methoxyisonicotinaldehyde ≥97% research grade
5-Chloro-2-methoxyisonicotinaldehyde


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856042
CAS Number: 1060810-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty