Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-4-formylbenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

17913-1G 17913-25G 17913-500MG 17913-5G
76926-312EA 126.25 USD InStock
76926-312 76926-314 76926-316 76926-318
3,5-Difluoro-4-formylbenzoic acid ≥95%
3,5-Difluoro-4-formylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₄F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09835188
CAS Number: 736990-88-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty