Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Cyclopropylpyridin-3-ylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

18314-1G 18314-25G 18314-500MG 18314-5G
76926-436EA 260.99 USD InStock
76926-436 76926-438 76926-440 76926-442
5-Cyclopropylpyridin-3-ylboronic acid ≥97%
5-Cyclopropylpyridin-3-ylboronic acid


Formula: C₈H₁₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11870259
CAS Number: 1044210-58-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty