Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,3-difluorophenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

18381-1G 18381-25G 18381-5G
76926-470EA 114.58 USD InStock
76926-470 76926-472 76926-474
4-Chloro-2,3-difluorophenylboronic acid ≥97% research grade
4-Chloro-2,3-difluorophenylboronic acid


Formula: C₆H₄BClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16660056
CAS Number: 1160561-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty