Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-methoxyphenol benzyl ether ≥97%

Supplier: AOB CHEM USA
Warning

19396-1G 19396-25G 19396-500MG 19396-5G
76926-764EA 192.03 USD InStock
76926-764 76926-766 76926-768 76926-770
4-Bromo-3-methoxyphenol benzyl ether ≥97%
4-Bromo-3-methoxyphenol benzyl ether
Precaución: for Research Use Only


Formula: C₁₄H₁₃BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02258915
CAS Number: 171768-67-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty