Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formyl-2-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

19513-1G 19513-25G 19513-500MG 19513-5G
76926-778EA 153.84 USD InStock
76926-778 76926-780 76926-782 76926-784
5-Formyl-2-methylbenzoic acid ≥97%
5-Formyl-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD26673176
CAS Number: 105650-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty