Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-hydroxypyridine ≥97%

Supplier: AOB CHEM USA
Warning

20459-1G 20459-25G 20459-500MG 20459-5G
76927-096EA 290.69 USD InStock
76927-096 76927-098 76927-100 76927-102
4-Bromo-2-fluoro-5-hydroxypyridine ≥97%
4-Bromo-2-fluoro-5-hydroxypyridine
Precaución: for Research Use Only


Formula: C₅H₃BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD16609914
CAS Number: 1227578-23-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty