Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20583-1G 20583-25G 20583-500MG 20583-5G
76927-146EA 118.83 USD InStock
76927-146 76927-148 76927-150 76927-152
5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid ≥97% research grade
5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid


Formula: C₁₃H₁₇BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08235096
CAS Number: 874289-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty