Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Carbamoyl-2-fluorophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

20586-1G 20586-25G 20586-500MG 20586-5G
76927-162EA 126.25 USD InStock
76927-162 76927-164 76927-166 76927-168
5-Carbamoyl-2-fluorophenylboronic acid ≥97%
5-Carbamoyl-2-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08436055
CAS Number: 874289-39-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty