Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluoronicotinic acid methyl ester ≥97%

Supplier: AOB CHEM USA
Warning

21051-1G 21051-25G 21051-500MG 21051-5G
76927-314EA 159.14 USD InStock
76927-314 76927-316 76927-318 76927-320
5-Chloro-2-fluoronicotinic acid methyl ester ≥97%
5-Chloro-2-fluoronicotinic acid methyl ester
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14635864
CAS Number: 1214324-17-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty