Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyclohexylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

22293-1G 22293-25G 22293-5G
76927-658EA 94.43 USD InStock
76927-658 76927-660 76927-662
4-Cyclohexylphenylboronic acid ≥97%
4-Cyclohexylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₇BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02093068
CAS Number: 374538-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty