Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-formylbenzoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

22609-1G 22609-25G 22609-5G
76927-770EA 76.39 USD InStock
76927-770 76927-772 76927-774
2-Fluoro-4-formylbenzoic acid ≥95% research grade
2-Fluoro-4-formylbenzoic acid


Formula: C₈H₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD12026481
CAS Number: 604000-97-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty