Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carboxy-2-chloro-3-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

22734-1G 22734-25G 22734-500MG 22734-5G
76927-806EA 231.28 USD InStock
76927-806 76927-808 76927-810 76927-812
4-Carboxy-2-chloro-3-methylphenylboronic acid ≥97%
4-Carboxy-2-chloro-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BClO₄
MW: 214.411 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526386
CAS Number: 1451391-31-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty