Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Ethoxymethyl)pyridine-3-boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

23497-1G 23497-500MG 23497-5G
76928-056EA 528.33 USD InStock
76928-056 76928-058 76928-060
5-(Ethoxymethyl)pyridine-3-boronic acid ≥97%
5-(Ethoxymethyl)pyridine-3-boronic acid
Precaución: for Research Use Only


Formula: C₈H₁₂BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11111800
CAS Number: 723281-60-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty