Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxypyridine-4-methanol ≥97%

Supplier: AOB CHEM USA
Warning

24355-1G 24355-25G 24355-500MG 24355-5G
76928-354EA 185.66 USD InStock
76928-354 76928-356 76928-358 76928-360
5-Bromo-2-methoxypyridine-4-methanol ≥97%
5-Bromo-2-methoxypyridine-4-methanol
Precaución: for Research Use Only


Formula: C₇H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16607017
CAS Number: 1227589-24-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty