Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-(trifluoromethyl)benzyl alcohol ≥97% research grade

Supplier: AOB CHEM USA
Warning

24555-1G 24555-25G 24555-5G
76928-432EA 100.79 USD InStock
76928-432 76928-434 76928-436
4-Bromo-3-(trifluoromethyl)benzyl alcohol ≥97% research grade
4-Bromo-3-(trifluoromethyl)benzyl alcohol


Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD08059129
CAS Number: 957207-09-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty