Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Cyclopropyl-2-flluoropyridine-3-carboxaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

24898-1G 24898-25G 24898-500MG 24898-5G
76928-578EA 381.93 USD InStock
76928-578 76928-580 76928-582 76928-584
5-Cyclopropyl-2-flluoropyridine-3-carboxaldehyde ≥97%
5-Cyclopropyl-2-flluoropyridine-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₉H₈FNO
MW: 165.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28977163
CAS Number: 2244107-70-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty