Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-3,5-dimethylbenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

25512-1G 25512-25G 25512-5G
76928-798EA 79.57 USD InStock
76928-798 76928-800 76928-802
Methyl 4-bromo-3,5-dimethylbenzoate ≥97% research grade
Methyl 4-bromo-3,5-dimethylbenzoate


Formula: C₁₀H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09833419
CAS Number: 432022-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty