Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxy-2,3-dimethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

25546-1G 25546-25G 25546-500MG 25546-5G
76928-820EA 201.58 USD InStock
76928-820 76928-822 76928-824 76928-826
4-Ethoxy-2,3-dimethylphenylboronic acid ≥97%
4-Ethoxy-2,3-dimethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD23713128
CAS Number: 1451391-67-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty