Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-ethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

26282-1G 26282-25G 26282-500MG 26282-5G
76929-150EA 142.17 USD InStock
76929-150 76929-152 76929-154 76929-156
4-Chloro-3-ethylbenzoic acid ≥97%
4-Chloro-3-ethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11111064
CAS Number: 42044-90-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty