Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Trifluoromethoxybiphenyl-2-carbaldehyde ≥95%

Supplier: AOB CHEM USA
Warning

26847-1G 26847-25G 26847-500MG 26847-5G
76929-320EA 290.69 USD InStock
76929-320 76929-322 76929-324 76929-326
4'-Trifluoromethoxybiphenyl-2-carbaldehyde ≥95%
4'-Trifluoromethoxybiphenyl-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₉F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06739448
CAS Number: 728919-13-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty