Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-chloro-2-fluorobenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

27503-1G 27503-25G 27503-500MG 27503-5G
76929-502EA 264.17 USD InStock
76929-502 76929-504 76929-506 76929-508
4-Bromo-3-chloro-2-fluorobenzaldehyde ≥97%
4-Bromo-3-chloro-2-fluorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃BrClFO
Storage Temperature: Ambient
MDL Number: MFCD26958726
CAS Number: 1696224-75-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty