Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Ethylthiophene-2-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

28376-1G 28376-25G 28376-500MG 28376-5G
76929-782EA 290.69 USD InStock
76929-782 76929-784 76929-786 76929-788
5-Ethylthiophene-2-boronic acid pinacol ester ≥97%
5-Ethylthiophene-2-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₉BO₂S
Storage Temperature: Ambient
MDL Number: MFCD12032489
CAS Number: 1447710-10-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty