Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-trifluoromethylbenzoic acid ethyl ester ≥97%

Supplier: AOB CHEM USA
Warning

28695-1G 28695-25G 28695-500MG 28695-5G
76929-884EA 144.29 USD InStock
76929-884 76929-886 76929-888 76929-890
4-Fluoro-3-trifluoromethylbenzoic acid ethyl ester ≥97%
4-Fluoro-3-trifluoromethylbenzoic acid ethyl ester
Precaución: for Research Use Only


Formula: C₁₀H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06204415
CAS Number: 67515-63-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty