Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-(4-fluorophenyl)pyridine ≥97%

Supplier: AOB CHEM USA
Warning

28865-1G 28865-25G 28865-500MG 28865-5G
76929-912EA 272.66 USD InStock
76929-912 76929-914 76929-916 76929-918
5-Bromo-2-(4-fluorophenyl)pyridine ≥97%
5-Bromo-2-(4-fluorophenyl)pyridine
Precaución: for Research Use Only


Formula: C₁₁H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD11100950
CAS Number: 463336-07-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty