Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Benzyloxy-2-methylpyridine ≥97%

Supplier: AOB CHEM USA
Warning

29841-1G 29841-25G 29841-500MG 29841-5G
76930-094EA 308.73 USD InStock
76930-094 76930-096 76930-098 76930-100
5-Benzyloxy-2-methylpyridine ≥97%
5-Benzyloxy-2-methylpyridine
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO
MW: 199.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15523327
CAS Number: 63793-98-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty