Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluoro-6-iodophenol ≥97%

Supplier: AOB CHEM USA
Warning

30458-1G 30458-25G 30458-500MG 30458-5G
76930-324EA 177.18 USD InStock
76930-324 76930-326 76930-328 76930-330
4-Chloro-2-fluoro-6-iodophenol ≥97%
4-Chloro-2-fluoro-6-iodophenol
Precaución: for Research Use Only


Formula: C₆H₃ClFIO
MW: 272.443 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32902576
CAS Number: 1192815-08-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty