Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-(trifluoromethyl)benzeneboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

30622-25G 30622-5G
76930-340EA 289.63 USD InStock
76930-340 76930-342
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid pinacol ester ≥97% research grade
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid pinacol ester


Formula: C₁₃H₁₅BF₄O₂
MW: 290.065 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878546
CAS Number: 445303-14-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty