Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-(difluoromethyl)-2-methoxypyridine ≥95%

Supplier: AOB CHEM USA
Warning

30883-1G 30883-25G 30883-500MG 30883-5G
76930-464EA 233.4 USD InStock
76930-464 76930-466 76930-468 76930-470
5-Bromo-3-(difluoromethyl)-2-methoxypyridine ≥95%
5-Bromo-3-(difluoromethyl)-2-methoxypyridine


Formula: C₇H₆BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD23163928
CAS Number: 1254123-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty