Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,4-difluorophenol ≥97%

Supplier: AOB CHEM USA
Warning

31715-1G 31715-25G 31715-500MG 31715-5G
76930-724EA 192.03 USD InStock
76930-724 76930-726 76930-728 76930-730
5-Bromo-2,4-difluorophenol ≥97%
5-Bromo-2,4-difluorophenol


Formula: C₆H₃BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD20231458
CAS Number: 355423-48-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty