Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-5-iodobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

32139-1G 32139-25G 32139-500MG 32139-5G
76930-794EA 393.6 USD InStock
76930-794 76930-796 76930-798 76930-800
4-Bromo-2-fluoro-5-iodobenzoic acid ≥97% research grade
4-Bromo-2-fluoro-5-iodobenzoic acid


Formula: C₇H₃BrFIO₂
MW: 344.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107175
CAS Number: 1521585-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty