Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-ethylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

32385-1G 32385-25G 32385-500MG 32385-5G
76930-906EA 146.41 USD InStock
76930-906 76930-908 76930-910 76930-912
4-Bromo-2-ethylphenylboronic acid pinacol ester ≥97%
4-Bromo-2-ethylphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₀BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD29094700
CAS Number: 1674364-30-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty