Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-chloro-2-ethoxy-3-fluorobenzene ≥95%

Supplier: AOB CHEM USA
Warning

32603-1G 32603-25G 32603-500MG 32603-5G
76930-978EA 168.69 USD InStock
76930-978 76930-980 76930-982 76930-984
5-Bromo-1-chloro-2-ethoxy-3-fluorobenzene ≥95%
5-Bromo-1-chloro-2-ethoxy-3-fluorobenzene
Precaución: for Research Use Only


Formula: C₈H₇BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28396344
CAS Number: 1936177-81-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty